Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ect2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195036.1 | NP_001181965.1 | 2 - 3 | 257 | 89 | |
GenBank mRNA | AK021214.1 | - | 2 - 3 | 258 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Mus musculus 1700 Probe spans exons |