Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamts9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175314.3 | NP_780523.2 | 5 - 6 | 648 | 68 | |
GenBank mRNA | CN534419.1 | - | 5 - 6 | 647 | 68 | |
CN703716.1 | - | 3 - 4 | 352 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Detect Genomic DNA Mus musculus Probe spans exons |