Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfxn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053196.3 | NP_444426.3 | 9 - 10 | 883 | 79 | |
GenBank mRNA | AF325261.1 | - | 9 - 10 | 841 | 79 | |
AK034875.1 | - | 797 | 79 | |||
AK149277.1 | - | 2715 | 79 | |||
AK150722.1 | - | 9 - 10 | 834 | 79 | ||
AK162407.1 | - | 7 - 8 | 2927 | 79 | ||
BC019808.1 | - | 9 - 10 | 881 | 5043343 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |