Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009315.3 | NP_033341.1 | 8 - 9 | 875 | 69 | |
GenBank mRNA | AK153731.1 | - | 923 | 69 | ||
AK154867.1 | - | 8 - 9 | 909 | 69 | ||
BC058583.1 | - | 8 - 9 | 875 | 6514960 | 69 | |
D49439.1 | - | 7 - 8 | 808 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |