Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500011245.mm.1 |
TC0500001288.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500011245.mm.1 |
NM_001081308.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500011245.mm.1 |
NM_001081308.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500011245.mm.1 |
NM_001199685.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500011245.mm.1 |
NM_001199685.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500011245.mm.1 |
NM_183306.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500011245.mm.1 |
NM_183306.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500011244.mm.1 |
TC0500001288.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500011244.mm.1 |
NM_001081308.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500011244.mm.1 |
NM_001081308.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500011244.mm.1 |
NM_001199685.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500011244.mm.1 |
NM_001199685.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500011244.mm.1 |
NM_183306.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500011244.mm.1 |
NM_183306.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500011243.mm.1 |
TC0500001288.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500011243.mm.1 |
NM_001081308.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500011243.mm.1 |
NM_001081308.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500011243.mm.1 |
NM_001199685.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500011243.mm.1 |
NM_001199685.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500011243.mm.1 |
NM_183306.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500011243.mm.1 |
NM_183306.2 |
Refseq |