Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK003572.2 | - | 3 - 4 | 219 | 65 | |
AK085669.1 | - | 3 - 4 | 268 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |