Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfkb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177369.1 | NP_001170840.1 | 2 - 3 | 180 | 124 | |
NM_001177370.1 | NP_001170841.1 | 1 - 2 | 96 | 124 | ||
NM_019408.3 | NP_062281.1 | 1 - 2 | 331 | 124 | ||
GenBank mRNA | AF155372.1 | - | 2 - 3 | 258 | 124 | |
AF155373.1 | - | 2 - 3 | 295 | 124 | ||
AK134081.1 | - | 2 - 3 | 598 | 125 | ||
AK137680.1 | - | 2 - 3 | 170 | 124 | ||
AK148160.1 | - | 2 - 3 | 168 | 124 | ||
AK150012.1 | - | 2 - 3 | 159 | 124 | ||
BC027423.1 | - | 1 - 2 | 97 | 3672913 | 124 | |
CJ134893.1 | - | 1 - 2 | 328 | 124 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |