Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfxn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001178013.1 | NP_001171484.1 | 4 - 5 | 753 | 136 | |
NM_053197.4 | NP_444427.1 | 4 - 5 | 756 | 136 | ||
GenBank mRNA | AF325262.1 | - | 4 - 5 | 520 | 136 | |
AK034514.1 | - | 4 - 5 | 473 | 136 | ||
AK154274.1 | - | 4 - 5 | 526 | 136 | ||
BC012208.1 | - | 4 - 5 | 658 | 3985155 | 136 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |