Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsc22d4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029805.1 | NP_084081.1 | 5 - 6 | 2000 | 114 | |
GenBank mRNA | AK155712.1 | - | 5 - 6 | 2002 | 114 | |
AK170487.1 | - | 5 - 6 | 1989 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mus musculus 1700 Probe spans exons |