Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Morn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198108.2 | NP_932776.1 | 2 - 3 | 277 | 100 | |
GenBank mRNA | AK154147.1 | - | 2 - 3 | 265 | 100 | |
AK159230.1 | - | 2 - 3 | 264 | 100 | ||
BB871690.1 | - | 2 - 3 | 100 | |||
BC023055.1 | - | 2 - 3 | 244 | 100 | ||
BC057060.1 | - | 1 - 2 | 653 | 100 | ||
CV563048.1 | - | 2 - 3 | 203 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |