Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cngb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013927.2 | NP_038955.1 | 9 - 10 | 1116 | 77 | |
GenBank mRNA | AJ243572.1 | - | 9 - 10 | 1116 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Mus musculus Probe spans exons |