Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008872.2 | NP_032898.2 | 9 - 10 | 995 | 101 | |
GenBank mRNA | AK135857.1 | - | 9 - 10 | 986 | 101 | |
AK135965.1 | - | 9 - 10 | 987 | 101 | ||
AK141665.1 | - | 9 - 10 | 984 | 101 | ||
AK159620.1 | - | 9 - 10 | 987 | 101 | ||
AK166639.1 | - | 9 - 10 | 985 | 101 | ||
BC011256.1 | - | 9 - 10 | 992 | 4038404 | 101 | |
BC057967.1 | - | 9 - 10 | 975 | 4979664 | 101 | |
BC061508.1 | - | 9 - 10 | 976 | 5352018 | 101 | |
J03520.1 | - | 9 - 10 | 978 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Mus musculus Probe spans exons |