Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026767.4 | NP_081043.1 | 1 - 2 | 146 | 65 | |
GenBank mRNA | AK003223.1 | - | 1 - 2 | 56 | 65 | |
CJ237809.1 | - | 1 - 2 | 146 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |