Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrna6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021369.2 | NP_067344.2 | 4 - 5 | 618 | 61 | |
GenBank mRNA | AJ245706.1 | - | 4 - 5 | 456 | 61 | |
AK149262.1 | - | 4 - 5 | 559 | 61 | ||
AY574265.1 | - | 4 - 5 | 376 | 61 | ||
BC031985.1 | - | 4 - 5 | 618 | 4501558 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons |