Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003933.2 | NP_001003933.1 | 6 - 7 | 2936 | 69 | |
NM_001003934.2 | NP_001003934.1 | 7 - 8 | 2993 | 69 | ||
NM_001271486.1 | NP_001258415.1 | 7 - 8 | 2030 | 69 | ||
NM_001271487.1 | NP_001258416.1 | 6 - 7 | 869 | 69 | ||
NM_053076.3 | NP_444306.1 | 5 - 6 | 812 | 69 | ||
GenBank mRNA | AB046114.1 | - | 5 - 6 | 806 | 69 | |
AF195940.1 | - | 5 - 6 | 756 | 69 | ||
AK032109.1 | - | 7 - 8 | 2007 | 69 | ||
AK036892.1 | - | 5 - 6 | 778 | 69 | ||
AK049845.1 | - | 5 - 6 | 744 | 69 | ||
AK075883.1 | - | 5 - 6 | 741 | 69 | ||
AK088670.1 | - | 5 - 6 | 744 | 69 | ||
AK146505.1 | - | 5 - 6 | 620 | 69 | ||
AK148792.1 | - | 5 - 6 | 771 | 69 | ||
AK148947.1 | - | 5 - 6 | 773 | 69 | ||
AY164700.1 | - | 5 - 6 | 806 | 69 | ||
AY427822.1 | - | 6 - 7 | 2917 | 69 | ||
AY507126.1 | - | 5 - 6 | 604 | 69 | ||
AY507127.1 | - | 5 - 6 | 2539 | 69 | ||
AY750849.1 | - | 7 - 8 | 2974 | 69 | ||
BC005478.1 | - | 5 - 6 | 715 | 69 | ||
BC014697.1 | - | 5 - 6 | 723 | 69 | ||
BC036717.1 | - | 5 - 6 | 721 | 69 | ||
BC079882.1 | - | 7 - 8 | 2903 | 6839323 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |