Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ldb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077398.2 | NP_001070866.1 | 1 - 2 | 390 | 85 | |
NM_001286348.1 | NP_001273277.1 | 1 - 2 | 390 | 85 | ||
NM_010698.4 | NP_034828.3 | 1 - 2 | 390 | 85 | ||
GenBank mRNA | AB221652.1 | - | 1 - 2 | 137 | 85 | |
AK146860.1 | - | 1 - 2 | 324 | 85 | ||
AK147160.1 | - | 1 - 2 | 300 | 85 | ||
BC079611.1 | - | 1 - 2 | 283 | 30360903 | 85 | |
CN670130.1 | - | 1 - 2 | 352 | 85 | ||
U89487.1 | - | 1 - 2 | 279 | 85 | ||
U89489.1 | - | 1 - 2 | 279 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |