Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcl11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016707.3 | NP_057916.1 | 4 - 5 | 2565 | 101 | |
GenBank mRNA | AF051525.1 | - | 4 - 5 | 2503 | 101 | |
AF169036.1 | - | 2 - 3 | 1706 | 101 | ||
BC010585.1 | - | 1 - 2 | 1579 | 2631265 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |