Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Socs5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019654.2 | NP_062628.2 | 1 - 2 | 366 | 64 | |
GenBank mRNA | AK048342.1 | - | 1 - 2 | 187 | 64 | |
BC053015.1 | - | 1 - 2 | 366 | 5702321 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |