Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thoc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153552.3 | NP_705780.1 | 14 - 15 | 1146 | 96 | |
GenBank mRNA | AK031785.1 | - | 14 - 15 | 1147 | 96 | |
AK032200.1 | - | 14 - 15 | 1147 | 96 | ||
AK052835.1 | - | 14 - 15 | 1142 | 96 | ||
AK167374.1 | - | 14 - 15 | 1160 | 96 | ||
AK168619.1 | - | 14 - 15 | 1412 | 96 | ||
BC024951.1 | - | 14 - 15 | 1318 | 4010605 | 96 | |
BC027149.1 | - | 7 - 8 | 925 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |