Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccny | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026484.3 | NP_080760.2 | 8 - 9 | 1222 | 102 | |
GenBank mRNA | AK017493.1 | - | 8 - 9 | 1054 | 102 | |
AK028696.1 | - | 8 - 9 | 1223 | 102 | ||
AK076556.1 | - | 8 - 9 | 752 | 102 | ||
BC023321.1 | - | 9 - 10 | 1140 | 3995040 | 102 | |
BC035524.1 | - | 7 - 8 | 977 | 4974858 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |