Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Otx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011023.3 | NP_035153.1 | 1 - 2 | 106 | 71 | |
GenBank mRNA | AK042975.1 | - | 1 - 2 | 70 | 71 | |
AK049430.1 | - | 1 - 2 | 166 | 71 | ||
AK050758.1 | - | 1 - 2 | 67 | 71 | ||
AK083434.1 | - | 1 - 2 | 68 | 71 | ||
AK083671.1 | - | 1 - 2 | 50 | 71 | ||
AK158588.1 | - | 1 - 2 | 71 | 71 | ||
BB652255.1 | - | 1 - 2 | 164 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus 1700 Probe spans exons |