Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Peli1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023324.2 | NP_075813.2 | 2 - 3 | 487 | 82 | |
GenBank mRNA | AF302503.1 | - | 1 - 2 | 709 | 82 | |
AK020915.1 | - | 2 - 3 | 521 | 82 | ||
AK029586.1 | - | 2 - 3 | 385 | 82 | ||
AK151660.1 | - | 2 - 3 | 488 | 82 | ||
AK152900.1 | - | 2 - 3 | 488 | 82 | ||
AK152997.1 | - | 2 - 3 | 488 | 82 | ||
AK153372.1 | - | 2 - 3 | 488 | 82 | ||
AK167525.1 | - | 2 - 3 | 488 | 82 | ||
BC016515.1 | - | 1 - 2 | 86 | 3600130 | 82 | |
BC026695.1 | - | 2 - 3 | 451 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |