Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctsf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019861.1 | NP_063914.1 | 9 - 10 | 1120 | 66 | |
GenBank mRNA | AF136280.1 | - | 9 - 10 | 1021 | 66 | |
AJ131851.1 | - | 9 - 10 | 997 | 66 | ||
AK017474.1 | - | 9 - 10 | 1003 | 66 | ||
AK075862.1 | - | 9 - 10 | 1013 | 66 | ||
AK207816.1 | - | 2 - 3 | 139 | 66 | ||
BC004054.1 | - | 6 - 7 | 591 | 66 | ||
BC051131.1 | - | 8 - 9 | 1239 | 66 | ||
BC058758.1 | - | 9 - 10 | 1011 | 6492834 | 66 | |
BC115438.1 | - | 3 - 4 | 240 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Mus musculus Probe spans exons |