Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdzd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133226.2 | NP_573489.2 | 2 - 3 | 48 | 71 | |
GenBank mRNA | AF334612.1 | - | 2 - 3 | 44 | 71 | |
AK052748.1 | - | 2 - 3 | 49 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |