Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172117.1 | NP_001165588.1 | 6 - 7 | 752 | 98 | |
NM_010407.4 | NP_034537.2 | 6 - 7 | 752 | 98 | ||
GenBank mRNA | AK149736.1 | - | 6 - 7 | 718 | 98 | |
AK150290.1 | - | 6 - 7 | 726 | 98 | ||
AK150709.1 | - | 6 - 7 | 735 | 98 | ||
AK152506.1 | - | 6 - 7 | 726 | 98 | ||
AK155975.1 | - | 6 - 7 | 665 | 98 | ||
AK159234.1 | - | 6 - 7 | 685 | 98 | ||
AK165315.1 | - | 6 - 7 | 717 | 98 | ||
BC010478.1 | - | 6 - 7 | 748 | 3978518 | 98 | |
J03023.1 | - | 6 - 7 | 641 | 98 | ||
Y00487.1 | - | 6 - 7 | 647 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |