Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tlk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112705.2 | NP_001106176.1 | 14 - 15 | 1816 | 134 | |
NM_001294329.1 | NP_001281258.1 | 12 - 13 | 1676 | 134 | ||
NM_001294330.1 | NP_001281259.1 | 12 - 13 | 1624 | 134 | ||
NM_001294331.1 | NP_001281260.1 | 14 - 15 | 1829 | 134 | ||
NM_001294334.1 | NP_001281263.1 | 14 - 15 | 1816 | 134 | ||
GenBank mRNA | AF045252.1 | - | 14 - 15 | 1835 | 134 | |
AF045253.1 | - | 13 - 14 | 1263 | 134 | ||
AF045254.1 | - | 9 - 10 | 977 | 134 | ||
AK014829.1 | - | 14 - 15 | 1454 | 134 | ||
AK089925.1 | - | 12 - 13 | 1204 | 134 | ||
AK165667.1 | - | 14 - 15 | 1819 | 134 | ||
AX775743.1 | - | 8 - 9 | 841 | 134 | ||
BC066198.1 | - | 12 - 13 | 1169 | 134 | ||
U56909.1 | - | 5 - 6 | 514 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |