Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153103.2 | NP_694743.2 | 21 - 22 | 2382 | 114 | |
GenBank mRNA | AB074017.1 | - | 19 - 20 | 2012 | 114 | |
AK046574.1 | - | 16 - 17 | 1623 | 114 | ||
BC006188.1 | - | 3 - 4 | 330 | 114 | ||
BC029286.1 | - | 21 - 22 | 2369 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |