Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mylk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081044.2 | NP_001074513.2 | 3 - 4 | 1016 | 84 | |
GenBank mRNA | AK079396.1 | - | 3 - 4 | 1021 | 84 | |
BC158041.1 | - | 3 - 4 | 847 | 9088157 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |