Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146254.4 | NP_666366.4 | 3 - 4 | 555 | 148 | |
GenBank mRNA | BC028975.1 | - | 3 - 4 | 480 | 5363117 | 148 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |