Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mier1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039081.2 | NP_001034170.1 | 9 - 10 | 951 | 78 | |
NM_001286221.1 | NP_001273150.1 | 11 - 12 | 1082 | 78 | ||
NM_001286222.1 | NP_001273151.1 | 11 - 12 | 1136 | 78 | ||
NM_001286223.1 | NP_001273152.1 | 9 - 10 | 932 | 78 | ||
NM_027696.3 | NP_081972.2 | 11 - 12 | 1139 | 78 | ||
GenBank mRNA | AK016915.1 | - | 3 - 4 | 381 | 78 | |
AK129405.1 | - | 11 - 12 | 1059 | 78 | ||
AY769435.1 | - | 10 - 11 | 931 | 78 | ||
AY769436.1 | - | 9 - 10 | 854 | 78 | ||
AY769437.1 | - | 9 - 10 | 904 | 78 | ||
AY769438.1 | - | 11 - 12 | 1004 | 78 | ||
BC108273.1 | - | 9 - 10 | 918 | 30080243 | 78 | |
BC130227.1 | - | 9 - 10 | 903 | 40141863 | 78 | |
CN538186.1 | - | 6 - 7 | 552 | 78 | ||
CN701986.1 | - | 3 - 4 | 305 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |