Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vil1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009509.2 | NP_033535.2 | 6 - 7 | 634 | 64 | |
GenBank mRNA | AK027908.1 | - | 6 - 7 | 631 | 64 | |
BC015267.1 | - | 6 - 7 | 610 | 4236751 | 64 | |
BC117875.2 | - | 6 - 7 | 599 | 40095718 | 64 | |
BC145671.1 | - | 6 - 7 | 601 | 40130974 | 64 | |
M98454.1 | - | 6 - 7 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |