Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp5a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007505.2 | NP_031531.1 | 1 - 2 | 343 | 101 | |
GenBank mRNA | AK043976.1 | - | 1 - 2 | 111 | 101 | |
AK076572.1 | - | 1 - 2 | 111 | 101 | ||
AK098155.1 | - | 1 - 2 | 121 | 101 | ||
AK146797.1 | - | 1 - 2 | 114 | 101 | ||
AK150426.1 | - | 1 - 2 | 114 | 101 | ||
AK150843.1 | - | 1 - 2 | 111 | 101 | ||
AK151004.1 | - | 1 - 2 | 111 | 101 | ||
AK151128.1 | - | 1 - 2 | 120 | 101 | ||
AK151224.1 | - | 1 - 2 | 118 | 101 | ||
AK151920.1 | - | 1 - 2 | 115 | 101 | ||
AK152054.1 | - | 1 - 2 | 122 | 101 | ||
AK152890.1 | - | 1 - 2 | 114 | 101 | ||
AK153167.1 | - | 1 - 2 | 115 | 101 | ||
AK153282.1 | - | 1 - 2 | 111 | 101 | ||
AK159491.1 | - | 1 - 2 | 111 | 101 | ||
AK159540.1 | - | 1 - 2 | 111 | 101 | ||
AK160043.1 | - | 1 - 2 | 121 | 101 | ||
AK166709.1 | - | 1 - 2 | 114 | 101 | ||
AK166812.1 | - | 1 - 2 | 127 | 101 | ||
AK167159.1 | - | 1 - 2 | 111 | 101 | ||
AK167863.1 | - | 1 - 2 | 91 | 101 | ||
AK168198.1 | - | 1 - 2 | 121 | 101 | ||
AK168369.1 | - | 1 - 2 | 111 | 101 | ||
AK168617.1 | - | 1 - 2 | 111 | 101 | ||
AK168879.1 | - | 1 - 2 | 111 | 101 | ||
AK168890.1 | - | 1 - 2 | 111 | 101 | ||
AK168932.1 | - | 1 - 2 | 111 | 101 | ||
AK169080.1 | - | 1 - 2 | 114 | 101 | ||
AK169084.1 | - | 1 - 2 | 114 | 101 | ||
AK169105.1 | - | 1 - 2 | 111 | 101 | ||
AK169142.1 | - | 1 - 2 | 156 | 101 | ||
AK169300.1 | - | 1 - 2 | 111 | 101 | ||
AK169308.1 | - | 1 - 2 | 116 | 101 | ||
AK169414.1 | - | 1 - 2 | 114 | 101 | ||
BC014854.1 | - | 1 - 2 | 98 | 4500893 | 101 | |
BC020417.1 | - | 1 - 2 | 98 | 101 | ||
CJ155200.1 | - | 1 - 2 | 344 | 101 | ||
L01062.1 | - | 1 - 2 | 98 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Transporter Mammalian Gene Collection Mus musculus 1700 Probe spans exons |