Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acsl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027976.2 | NP_082252.1 | 17 - 18 | 2417 | 111 | |
GenBank mRNA | AK006541.1 | - | 6 - 7 | 894 | 111 | |
AK034942.1 | - | 2417 | 111 | |||
AK145894.1 | - | 17 - 18 | 2078 | 111 | ||
AK150542.1 | - | 17 - 18 | 2083 | 111 | ||
AK150663.1 | - | 17 - 18 | 2083 | 111 | ||
AK151378.1 | - | 17 - 18 | 1872 | 111 | ||
AK161024.1 | - | 17 - 18 | 1770 | 111 | ||
AK168019.1 | - | 5 - 6 | 418 | 111 | ||
BC031544.1 | - | 17 - 18 | 1851 | 3987201 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |