Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205236.1 | NP_001192165.1 | 16 - 17 | 2898 | 111 | |
NM_133715.5 | NP_598476.2 | 14 - 15 | 1867 | 111 | ||
GenBank mRNA | AK046763.1 | - | 14 - 15 | 1868 | 111 | |
AK143916.1 | - | 12 - 13 | 1339 | 111 | ||
AK154926.1 | - | 15 - 16 | 3189 | 111 | ||
BC096671.1 | - | 1 - 2 | 93 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |