Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grpel2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021296.2 | NP_067271.1 | 2 - 3 | 257 | 75 | |
GenBank mRNA | AF041060.1 | - | 2 - 3 | 222 | 75 | |
AK003011.1 | - | 2 - 3 | 238 | 75 | ||
AK012634.1 | - | 2 - 3 | 236 | 75 | ||
AK082983.1 | - | 247 | 75 | |||
AK134455.1 | - | 2 - 3 | 236 | 75 | ||
AK145493.1 | - | 2 - 3 | 249 | 75 | ||
BC068232.1 | - | 2 - 3 | 234 | 30533079 | 75 | |
BC094583.1 | - | 2 - 3 | 224 | 75 | ||
BC120747.1 | - | 2 - 3 | 247 | 40129670 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |