Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003917.4 | NP_001003917.2 | 3 - 4 | 375 | 79 | |
NM_001288612.1 | NP_001275541.1 | 3 - 4 | 378 | 79 | ||
NM_001288613.1 | NP_001275542.1 | 3 - 4 | 375 | 79 | ||
NR_109938.1 | 3 - 4 | 378 | 79 | |||
GenBank mRNA | AK043624.1 | - | 3 - 4 | 429 | 79 | |
AM085510.1 | - | 3 - 4 | 288 | 79 | ||
BC056488.1 | - | 3 - 4 | 288 | 79 | ||
BC079884.1 | - | 3 - 4 | 323 | 6840050 | 79 | |
BC082307.1 | - | 1 - 2 | 81 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |