Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St8sia3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009182.3 | NP_033208.2 | 1 - 2 | 328 | 81 | |
GenBank mRNA | AK015874.1 | - | 1 - 2 | 312 | 81 | |
AK161482.1 | - | 1 - 2 | 329 | 81 | ||
BB616760.1 | - | 1 - 2 | 329 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus 1700 Probe spans exons |