Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp2r5d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009358.3 | NP_033384.2 | 10 - 11 | 1373 | 158 | |
GenBank mRNA | AB053464.1 | - | 10 - 11 | 1318 | 158 | |
AF316616.2 | - | 10 - 11 | 1230 | 158 | ||
AK154050.1 | - | 10 - 11 | 1349 | 158 | ||
AK160587.1 | - | 10 - 11 | 1314 | 158 | ||
AK211926.1 | - | 2 - 3 | 191 | 158 | ||
AY338234.1 | - | 10 - 11 | 1303 | 158 | ||
BC010716.1 | - | 10 - 11 | 1286 | 2655861 | 158 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Phosphatase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |