Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rsph9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029338.3 | NP_083614.1 | 2 - 3 | 440 | 65 | |
GenBank mRNA | AK006440.1 | - | 2 - 3 | 443 | 65 | |
AK165820.1 | - | 2 - 3 | 311 | 65 | ||
BC019423.1 | - | 2 - 3 | 414 | 4922914 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |