Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082552.2 | NP_001076021.1 | 1 - 2 | 95 | 95 | |
GenBank mRNA | AK150484.1 | - | 1 - 2 | 59 | 95 | |
AK152619.1 | - | 1 - 2 | 59 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |