Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145832.3 | NP_665831.1 | 1 - 2 | 118 | 118 | |
GenBank mRNA | AK045972.1 | - | 1 - 2 | 95 | 118 | |
AK050311.1 | - | 1 - 2 | 83 | 118 | ||
AK088227.1 | - | 1 - 2 | 105 | 118 | ||
BY021673.1 | - | 1 - 2 | 138 | 118 | ||
BY026060.1 | - | 1 - 2 | 119 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mus musculus 1700 Probe spans exons |