Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnpla2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163689.1 | NP_001157161.1 | 4 - 5 | 796 | 76 | |
NM_025802.3 | NP_080078.2 | 4 - 5 | 796 | 76 | ||
NR_028142.1 | 5 - 6 | 878 | 76 | |||
GenBank mRNA | AK002826.1 | - | 4 - 5 | 778 | 76 | |
AK031609.1 | - | 4 - 5 | 754 | 76 | ||
AK076464.1 | - | 4 - 5 | 580 | 76 | ||
AK085926.1 | - | 4 - 5 | 781 | 76 | ||
AK150184.1 | - | 4 - 5 | 778 | 76 | ||
AY510273.1 | - | 4 - 5 | 777 | 76 | ||
AY731699.1 | - | 4 - 5 | 777 | 76 | ||
AY894805.1 | - | 4 - 5 | 697 | 76 | ||
BC019188.1 | - | 5 - 6 | 860 | 5027544 | 76 | |
BC034190.1 | - | 3 - 4 | 297 | 76 | ||
BC044737.1 | - | 5 - 6 | 842 | 76 | ||
BC044781.2 | - | 5 - 6 | 842 | 5151911 | 76 | |
BC054825.1 | - | 5 - 6 | 758 | 76 | ||
BC064747.1 | - | 4 - 5 | 768 | 30024535 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |