Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsmce4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162855.1 | NP_001156327.1 | 4 - 5 | 751 | 86 | |
GenBank mRNA | AK010349.1 | - | 4 - 5 | 751 | 86 | |
AK010795.1 | - | 4 - 5 | 717 | 86 | ||
AK179472.1 | - | 3 - 4 | 357 | 86 | ||
BC032990.1 | - | 7 - 8 | 86 | |||
BC051572.1 | - | 7 - 8 | 86 | |||
BC089629.1 | - | 1 - 2 | 109 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |