Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tjap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252473.1 | NP_001239402.1 | 3 - 4 | 252 | 75 | |
NM_028751.3 | NP_083027.1 | 4 - 5 | 382 | 75 | ||
GenBank mRNA | AK002882.2 | - | 4 - 5 | 369 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |