Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H2-M2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY302212.1 | - | 4 - 5 | 907 | 120 | |
AY302213.1 | - | 4 - 5 | 907 | 120 | ||
AY302214.1 | - | 4 - 5 | 907 | 120 | ||
AY302215.1 | - | 4 - 5 | 907 | 120 | ||
AY302216.1 | - | 4 - 5 | 907 | 120 | ||
BC144938.1 | - | 4 - 5 | 932 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons |