Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mccc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030026.2 | NP_084302.1 | 7 - 8 | 869 | 105 | |
GenBank mRNA | AK132265.1 | - | 7 - 8 | 849 | 105 | |
AK143233.1 | - | 7 - 8 | 870 | 105 | ||
AK145564.1 | - | 7 - 8 | 821 | 105 | ||
AK146844.1 | - | 7 - 8 | 829 | 105 | ||
AK146865.1 | - | 7 - 8 | 819 | 105 | ||
BC141193.1 | - | 7 - 8 | 835 | 9055805 | 105 | |
BC141194.1 | - | 7 - 8 | 835 | 9055806 | 105 | |
BQ551658.1 | - | 3 - 4 | 321 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |