Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cetn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145825.2 | NP_665824.1 | 4 - 5 | 502 | 80 | |
GenBank mRNA | AF362367.1 | - | 4 - 5 | 502 | 80 | |
AK078275.1 | - | 3 - 4 | 2417 | 80 | ||
AK090064.1 | - | 360 | 80 | |||
AK131808.1 | - | 4 - 5 | 491 | 80 | ||
BC038154.1 | - | 3 - 4 | 393 | 80 | ||
BC060991.1 | - | 4 - 5 | 454 | 6742836 | 80 | |
BC087905.1 | - | 4 - 5 | 480 | 6707456 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |