Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164056.1 | NP_001157528.1 | 14 - 15 | 1593 | 90 | |
NM_008875.4 | NP_032901.2 | 13 - 14 | 1538 | 90 | ||
GenBank mRNA | AK136111.1 | - | 8 - 9 | 776 | 90 | |
AK149229.1 | - | 1 - 2 | 1766 | 90 | ||
BC068144.1 | - | 13 - 14 | 1395 | 30532470 | 90 | |
U87868.1 | - | 14 - 15 | 1576 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |