Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3ca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008839.2 | NP_032865.2 | 3 - 4 | 867 | 67 | |
GenBank mRNA | AK051885.1 | - | 3 - 4 | 500 | 67 | |
BC089038.1 | - | 3 - 4 | 867 | 30692343 | 67 | |
BC130228.1 | - | 3 - 4 | 816 | 40141870 | 67 | |
U03279.1 | - | 3 - 4 | 812 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |