Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sctr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012322.2 | NP_001012322.2 | 9 - 10 | 1173 | 74 | |
GenBank mRNA | AK032652.1 | - | 9 - 10 | 1175 | 74 | |
AY255623.1 | - | 1 - 2 | 62 | 74 | ||
BC089355.1 | - | 8 - 9 | 1074 | 30551620 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |